Learn More
STEMCELL Technologies JAK Inhibitor I, Size: 1 mg
SDP
Catalog No. 501976166
Encompass
Janus Associated Kinase (JAK) Inhibitor I is a pyridine-containing tetracycle that disrupts JAK activity by interacting with the ATP binding domain. This inhibitor inhibits mouse JAK3 with Ki = 5 nM, JAK family members TYK2 and JAK2 with IC50 = 1 nM, and mouse JAK1 with IC50 = 15 nM, while showing weaker inhibition of other kinases (Thompson et al.). REPROGRAMMING· Inhibits the generation of mouse induced pluripotent stem cells (Efe et al.; Kim et al.).IMMUNOLOGY· Inhibits anti-viral response associated with JAK/STAT pathway activation (Xu et al.).
Catalog No. 50-197-6166 Supplier STEMCELL Technologies Supplier No. 74024
Janus Associated Kinase (JAK) Inhibitor I is a pyridine-containing tetracycle that disrupts JAK activity by interacting with the ATP binding domain. This inhibitor inhibits mouse JAK3 with Ki = 5 nM, JAK family members TYK2 and JAK2 with IC50 = 1 nM, and mouse JAK1 with IC50 = 15 nM, while showing weaker inhibition of other kinases (Thompson et al.). REPROGRAMMING· Inhibits the generation of mouse induced pluripotent stem cells (Efe et al.; Kim et al.).IMMUNOLOGY· Inhibits anti-viral response associated with JAK/STAT pathway activation (Xu et al.).