Learn More
Cell Signaling Technology Jak Isoform Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502729474
Encompass
The Jak Isoform Antibody Sampler Kit provides an economical means to examine several Jak isoforms, including Jak1, Jak2, and Jak3. The kit contains sufficient primary and secondary antibodies to perform two Western blot experiments per primary antibody. Included primary antibodies are rabbit monoclonal antibodies against Jak1, Jak2, and Jak3, along with an HRP-linked anti-rabbit IgG secondary antibody.
  • Enables examination of Jak1, Jak2, and Jak3 isoforms
  • Provides enough material for two Western blot experiments per primary antibody
  • Includes validated monoclonal antibodies and HRP-linked secondary antibody
  • Optimized for Western blotting, immunoprecipitation, and immunohistochemistry applications
Catalog No. 50-272-9474 Supplier Cell Signaling Technology Supplier No. 9945T
The Jak Isoform Antibody Sampler Kit provides an economical means to examine several Jak isoforms, including Jak1, Jak2, and Jak3. The kit contains sufficient primary and secondary antibodies to perform two Western blot experiments per primary antibody. Included primary antibodies are rabbit monoclonal antibodies against Jak1, Jak2, and Jak3, along with an HRP-linked anti-rabbit IgG secondary antibody.
  • Enables examination of Jak1, Jak2, and Jak3 isoforms
  • Provides enough material for two Western blot experiments per primary antibody
  • Includes validated monoclonal antibodies and HRP-linked secondary antibody
  • Optimized for Western blotting, immunoprecipitation, and immunohistochemistry applications