Learn More
Sino Biological Recombinant Human FLT3 Protein (Y591-V592insVDFREYEYD F691L, GST Tag), Active
Catalog No. 502574865
Encompass_Preferred
Recombinant human FLT3 (Y591-V592insVDFREYEYD F691L) (571-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. This FLT3-ITD mutant has 9 amino acids “VDFREYEYD” repeated between the sites Y591 and V592 within the juxtamembrane domain.
Catalog No. 50-257-4865 Supplier Sino Biological Supplier No. F1212LG50
Recombinant human FLT3 (Y591-V592insVDFREYEYD F691L) (571-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. This FLT3-ITD mutant has 9 amino acids “VDFREYEYD” repeated between the sites Y591 and V592 within the juxtamembrane domain.