Learn More
Medchemexpress LLC Jak1-in-8 | 1973485-18-5 | 95.0% | 442.51 | 100 MG
SDP
Catalog No. 5000351511
Encompass_Preferred
JAK1-IN-8 is a potent JAK1 inhibitor (IC50<500 nM), identified as compound 28, and extracted from patent WO2016119700A1. It targets JAK1 with high potency and has varying inhibitory effects on JAK2 and JAK3.
  • Potent JAK1 inhibitor with IC50 <500 nM
  • Targets JAK2 with IC50 between 500-1000 nM
  • Targets JAK3 with IC50 >5000 nM
Catalog No. 50-003-51511 Supplier Medchemexpress LLC Supplier No. HY139423100MG
JAK1-IN-8 is a potent JAK1 inhibitor (IC50<500 nM), identified as compound 28, and extracted from patent WO2016119700A1. It targets JAK1 with high potency and has varying inhibitory effects on JAK2 and JAK3.
  • Potent JAK1 inhibitor with IC50 <500 nM
  • Targets JAK2 with IC50 between 500-1000 nM
  • Targets JAK3 with IC50 >5000 nM