Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JAK1-IN-8 | 1973485-18-5 | 95.0% | 442.51 | 50 MG
SDP

Catalog No. 5000351185
Encompass_Preferred

JAK1-IN-8 is a potent JAK1 inhibitor (IC50<500 nM), identified as compound 28 and extracted from patent WO2016119700A1. It is intended for research use only.

  • Potent JAK1 inhibitor (IC50<500 nM)
  • Compound 28 from patent WO2016119700A1
  • Formula: C22H23FN4O3S
  • Appearance: White to off-white solid
  • Solubility in DMSO: 125 mg/mL (282.48 mM)
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-51185 Supplier Medchemexpress LLC Supplier No. HY13942350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JAK1-IN-8 is a potent JAK1 inhibitor (IC50<500 nM), identified as compound 28 and extracted from patent WO2016119700A1. It is intended for research use only.

  • Potent JAK1 inhibitor (IC50<500 nM)
  • Compound 28 from patent WO2016119700A1
  • Formula: C22H23FN4O3S
  • Appearance: White to off-white solid
  • Solubility in DMSO: 125 mg/mL (282.48 mM)
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.