Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Jak1-in-8 | 1973485-18-5 | 95.0% | 442.51 g/mol | C22H23FN4O3S | 5 MG
SDP

Catalog No. 5000221016
Encompass_Preferred

JAK1-IN-8 is a small-molecule Janus kinase 1 (JAK1) inhibitor for research use. It is described in patent literature and characterized for in vitro studies with reported JAK1 inhibitory activity (IC50 ≈500 nM). The compound is supplied as a white to off-white solid with 95.0% reported purity and is soluble in DMSO.

  • Reported JAK1 inhibitory activity (IC50 ≈500 nM)
  • Molecular formula: C22H23FN4O3S
  • Molecular weight: 442.51 g/mol
  • Purity: 95.0%
  • Appearance: white to off-white solid
  • Solubility: DMSO soluble (≈125 mg/mL); may require ultrasonic
  • Storage guidance: powder -20°C recommended for long-term storage

Catalog No. 50-002-21016 Supplier Medchemexpress LLC Supplier No. HY1394235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JAK1-IN-8 is a small-molecule Janus kinase 1 (JAK1) inhibitor for research use. It is described in patent literature and characterized for in vitro studies with reported JAK1 inhibitory activity (IC50 ≈500 nM). The compound is supplied as a white to off-white solid with 95.0% reported purity and is soluble in DMSO.

  • Reported JAK1 inhibitory activity (IC50 ≈500 nM)
  • Molecular formula: C22H23FN4O3S
  • Molecular weight: 442.51 g/mol
  • Purity: 95.0%
  • Appearance: white to off-white solid
  • Solubility: DMSO soluble (≈125 mg/mL); may require ultrasonic
  • Storage guidance: powder -20°C recommended for long-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.