Learn More
TARGETMOL CHEMICALS INC JAK1-IN-8 5MG
SDP
Catalog No. 502580205
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JAK1-IN-8 a specific inhibitor of Janus kinase 1 (JAK1 IC50<500 nM). purity: 98%
Catalog No. 50-258-0205 Supplier TARGETMOL CHEMICALS INC Supplier No. T358995MG
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JAK1-IN-8 a specific inhibitor of Janus kinase 1 (JAK1 IC50<500 nM). purity: 98%