Learn More
BPS Bioscience Inc JAK1(JANUS KINASE1)ASSAY,96RXN
SDP
Catalog No. NC2292079
Encompass
JAK1 (Janus Kinase 1) Assay Kit96 reactionsanus kinase (JAK) is a family of intracellular, nonreceptor tyrosine kinases that play an important role in cytokine-mediated signaling. This family includes four protein kinases; JAK1, JAK2, JAK3 and Tyk2. Various studies have suggested that regulation of JAK activity should be a promising approach to combat numerous diseases such as immune deficiencies and cancers. The JAK1 (Janus Kinase 1) Assay Kit is designed to measure JAK1 activity for screening and profiling applications using Kinase-Glo® MAX as a detection reagent.
This item is not returnable. View return policy
Catalog No. NC2292079 Supplier BPS Bioscience Inc Supplier No. 79518
JAK1 (Janus Kinase 1) Assay Kit96 reactionsanus kinase (JAK) is a family of intracellular, nonreceptor tyrosine kinases that play an important role in cytokine-mediated signaling. This family includes four protein kinases; JAK1, JAK2, JAK3 and Tyk2. Various studies have suggested that regulation of JAK activity should be a promising approach to combat numerous diseases such as immune deficiencies and cancers. The JAK1 (Janus Kinase 1) Assay Kit is designed to measure JAK1 activity for screening and profiling applications using Kinase-Glo® MAX as a detection reagent.