Learn More
Cayman Chemical JAK1humnrecombInt 10ug
SDP
Catalog No. 502949735
Encompass_Preferred
Source: Active recombinant human N-terminal GST-tagged JAK1 expressed in insect cells • Amino acids: 866-1,154 • MW: 60 kDa
Catalog No. 50-294-9735 Supplier Cayman Chemical Supplier No. 3374110 UG
Source: Active recombinant human N-terminal GST-tagged JAK1 expressed in insect cells • Amino acids: 866-1,154 • MW: 60 kDa