Learn More
Carnabio Usa Inc JAK2/10UG/CATALYTIC
Catalog No. 507540829
Encompass
JAK2, Catalytic domain, Wild type, Amino Acid 826-1132, NP_004963.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0829 Supplier Carnabio Usa Inc Supplier No. 0804510
JAK2, Catalytic domain, Wild type, Amino Acid 826-1132, NP_004963.1, Expressed in Insect (sf21); 10microg