Learn More
Carnabio Usa Inc JAK2/5UG/CATALYTIC
Catalog No. 507540472
Encompass
JAK2, Catalytic domain, Wild type, Amino Acid 826-1132, NP_004963.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0472 Supplier Carnabio Usa Inc Supplier No. 080455
JAK2, Catalytic domain, Wild type, Amino Acid 826-1132, NP_004963.1, Expressed in Insect (sf21); 5microg