Learn More
Eurofins Discoverx JAK2, active, 10 µg. (14-640)
Supplier:  Eurofins Discoverx 14640
Encompass
C-terminal 6xHis-tagged recombinant, human JAK2 amino acids 808-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9101