Learn More
Sino Biological Recombinant Human IRAK4 Protein (catalytic domain, GST Tag), Active
Catalog No. 502574640
Encompass_Preferred
Recombinant human IRAK4 (104-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4640 Supplier Sino Biological Supplier No. I1211G500
Recombinant human IRAK4 (104-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.