Learn More
Medchemexpress LLC Jak2 inhibitor v | 195371-52-9 | MFCD00023550 | 96.9% | 344.45 | C23H24N2O | 10 MG
SDP
Catalog No. 5000287147
Encompass_Preferred
NSC 42834 (JAK2 Inhibitor V) is a research-grade small-molecule inhibitor of Janus kinase 2 (JAK2). It inhibits autophosphorylation of wild-type JAK2 and the V617F mutant and is used in cellular and biochemical studies of JAK/STAT signaling and oncology-related research. The compound is supplied as a characterized solid with supporting analytical documentation.
  • Small-molecule JAK2 inhibitor active against JAK2-WT and JAK2-V617F.
  • Documented purity and analytical data provided.
  • Suitable for in vitro and preclinical research on JAK/STAT signaling.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Molecular weight 344.45 g/mol and formula C23H24N2O.
Catalog No. 50-002-87147 Supplier Medchemexpress LLC Supplier No. HY1548010MG
NSC 42834 (JAK2 Inhibitor V) is a research-grade small-molecule inhibitor of Janus kinase 2 (JAK2). It inhibits autophosphorylation of wild-type JAK2 and the V617F mutant and is used in cellular and biochemical studies of JAK/STAT signaling and oncology-related research. The compound is supplied as a characterized solid with supporting analytical documentation.
  • Small-molecule JAK2 inhibitor active against JAK2-WT and JAK2-V617F.
  • Documented purity and analytical data provided.
  • Suitable for in vitro and preclinical research on JAK/STAT signaling.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Molecular weight 344.45 g/mol and formula C23H24N2O.