Learn More
TARGETMOL CHEMICALS INC JAK2 INHIBITOR V 25MG
SDP
Catalog No. 502580417
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JAK2 Inhibitor V (JAK2 Inhibitor V Z3) is a novel specific inhibitor of Jak2 inhibiting Jak2-V617F and Jak2-WT autophosphorylation in a dose-dependent manner. purity: 99%
Catalog No. 50-258-0417 Supplier TARGETMOL CHEMICALS INC Supplier No. T304225MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JAK2 Inhibitor V (JAK2 Inhibitor V Z3) is a novel specific inhibitor of Jak2 inhibiting Jak2-V617F and Jak2-WT autophosphorylation in a dose-dependent manner. purity: 99%