Learn More
Cayman Chemical JAK2 Inhibitor V, Purity: ≥95%, CAS: 195371-52-9, 10 mg
SDP
Catalog No. 502980235
Encompass_Preferred
A selective inhibitor of the autophosphorylation of wild type and V617F mutant forms of JAK2, displaying IC50 values between 10 and 30 µM; blocks the proliferation of erythroleukemia cells and human hematopoietic progenitor cells expressing JAK2-V617F
Catalog No. 50-298-0235 Supplier Cayman Chemical Supplier No. 1768410 MG
A selective inhibitor of the autophosphorylation of wild type and V617F mutant forms of JAK2, displaying IC50 values between 10 and 30 µM; blocks the proliferation of erythroleukemia cells and human hematopoietic progenitor cells expressing JAK2-V617F