Learn More
Cayman Chemical JAK2 InhIbItr V 1mg
SDP
Catalog No. 502966104
Encompass_Preferred
A selective inhibitor of the autophosphorylation of wild type and V617F mutant forms of JAK2, displaying IC50 values between 10 and 30 µM; blocks the proliferation of erythroleukemia cells and human hematopoietic progenitor cells expressing JAK2-V617F
Catalog No. 50-296-6104 Supplier Cayman Chemical Supplier No. 176841 MG
Add to Cart
Add to Cart
A selective inhibitor of the autophosphorylation of wild type and V617F mutant forms of JAK2, displaying IC50 values between 10 and 30 µM; blocks the proliferation of erythroleukemia cells and human hematopoietic progenitor cells expressing JAK2-V617F