Learn More
Cayman Chemical JAK2 JH2 DomaInhumnrecom 10ug
SDP
Catalog No. 502943875
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged biotinylated JAK2 JH2 domain expressed in insect cells (Sf9) • Amino acids: 513-827 • MW: 39 kDa
Catalog No. 50-294-3875 Supplier Cayman Chemical Supplier No. 4227310 UG
Source: Active recombinant human C-terminal His-tagged biotinylated JAK2 JH2 domain expressed in insect cells (Sf9) • Amino acids: 513-827 • MW: 39 kDa