Learn More
Cayman Chemical JAK2 JH2 DomaInV617FW659A 20ug
SDP
Catalog No. 502943743
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged JAK2 (V617F, W659A, W777A, F794H mutant) JH2 domain expressed in insect cells (Sf9) • Amino acids: 536-812 • MW: 33 kDa
Catalog No. 50-294-3743 Supplier Cayman Chemical Supplier No. 4227420 UG
Source: Active recombinant human C-terminal His-tagged JAK2 (V617F, W659A, W777A, F794H mutant) JH2 domain expressed in insect cells (Sf9) • Amino acids: 536-812 • MW: 33 kDa