Learn More
Sino Biological Recombinant Human DGKI Protein (Full Length, His Tag), Active
Catalog No. 502575346
Encompass_Preferred
Recombinant full length human DGKI was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-5346 Supplier Sino Biological Supplier No. D2710BH500
Recombinant full length human DGKI was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.