Learn More
Cayman Chemical JAK2JH1ndJH2DomaInshum20ug
SDP
Catalog No. 502943405
Encompass_Preferred
Source: Active recombinant human N-terminal His- and GST-tagged JAK2 JH1 and JH2 domains expressed in insect cells (Sf9) • Amino acids: 532-1,132 • MW: 98 kDa
Catalog No. 50-294-3405 Supplier Cayman Chemical Supplier No. 4227220 UG
Source: Active recombinant human N-terminal His- and GST-tagged JAK2 JH1 and JH2 domains expressed in insect cells (Sf9) • Amino acids: 532-1,132 • MW: 98 kDa