Learn More
Medchemexpress LLC HY-19544 5mg Medchemexpress, JAK3-IN-1 CAS:1805787-93-2 Purity:>98%
SDP
Catalog No. 502082142
Encompass_Preferred
Medchemexpress, HY-19544 5mg JAK3-IN-1 CAS:1805787-93-2 JAK3-IN-1 is a potent, selective and orally active JAK3 inhibitor with an IC50 of 4.8 nM. JAK3-IN-1 shows over 180-fold more selective for JAK3 than JAK1 (IC50 of 896 nM) and JAK2 (IC50 of 1050 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2142 Supplier Medchemexpress LLC Supplier No. HY195445MG
Medchemexpress, HY-19544 5mg JAK3-IN-1 CAS:1805787-93-2 JAK3-IN-1 is a potent, selective and orally active JAK3 inhibitor with an IC50 of 4.8 nM. JAK3-IN-1 shows over 180-fold more selective for JAK3 than JAK1 (IC50 of 896 nM) and JAK2 (IC50 of 1050 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.