Learn More
TARGETMOL CHEMICALS INC JAK3-IN-14 5MG
SDP
Catalog No. 502579262
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JAK3-IN-14 is a potent selective and orally active FLT3 inhibitor with IC50s of ∼40 8 and 4 nM for FLT3-WT FLT3-D835Y and FLT3-D835H respectively. purity: 98%
Catalog No. 50-257-9262 Supplier TARGETMOL CHEMICALS INC Supplier No. T677545MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. JAK3-IN-14 is a potent selective and orally active FLT3 inhibitor with IC50s of ∼40 8 and 4 nM for FLT3-WT FLT3-D835Y and FLT3-D835H respectively. purity: 98%