Learn More
Medchemexpress LLC HY-101976 10mg Medchemexpress, JAK3-IN-6 CAS:1443235-95-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10197610MG
Encompass_Preferred
Medchemexpress, HY-101976 10mg JAK3-IN-6 CAS:1443235-95-7 JAK3-IN-6 is a potent, selective irreversible Janus Associated Kinase 3 (JAK3) inhibitor, with an IC50 of 0.15 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7942