Learn More
Medchemexpress LLC Jak3-in-6 | 1443235-95-7 | 99.9% | 350.37 | C19H18N4O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10197650MG
Encompass_Preferred
JAK3-IN-6 is a potent, selective, irreversible inhibitor of Janus associated kinase 3 (JAK3) with sub-nanomolar potency, provided as a solid for research use. It exhibits well-defined solubility and purity suitable for in vitro and in vivo experiments.
  • Potent, selective irreversible JAK3 inhibitor (IC50 0.15 nM).
  • High purity (99.9%).
  • Soluble in DMSO (100 mg/mL); in vivo formulations ≥2.17 mg/mL.
  • White to light yellow solid appearance.
  • Available in multiple pack sizes and DMSO solution formats.
  • Suitable for biochemical and cellular studies of JAK/STAT signaling.
Catalog No. 50-002-49189