Learn More
Cayman Chemical JAK3 JH2 DomaInhumnrecom 10ug
SDP
Catalog No. 502943285
Encompass_Preferred
Source: Active recombinant human C-terminal AVI- and His-tagged JAK3 JH2 domain expressed in insect cells (sf9) • Amino acids: 511-790 • MW: 34 kDa
Catalog No. 50-294-3285 Supplier Cayman Chemical Supplier No. 4227610 UG
Source: Active recombinant human C-terminal AVI- and His-tagged JAK3 JH2 domain expressed in insect cells (sf9) • Amino acids: 511-790 • MW: 34 kDa