Learn More
Aviva Systems Biology Corp Jakmip1 Antibody - N-terminal region (ARP41124_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41124_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Jakmip1. It was validated on Western Blot. . Alternate names: MGC125191, MGC125215ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-284-912