Learn More
ABclonal Technology JAKMIP2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A143871000UL
Encompass_Preferred
The protein encoded by this gene is reported to be a component of the Golgi matrix. It may act as a golgin protein by negatively regulating transit of secretory cargo and by acting as a structural scaffold of the Golgi. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-09096