Learn More
ABclonal Technology JAM-A/CD321/F11R Rabbit
SDP
Catalog No. 5000005639
Encompass_Preferred
Predicted to enable PDZ domain binding activity integrin binding activity and protein homodimerization activity Involved in intestinal absorption regulation of cytokine production and regulation of membrane permeability Acts upstream of or within cell adhesion and epithelial cell differentiation Located in bicellular tight junction Is expressed in several structures including 4-8 cell stage embryo alimentary system respiratory system sensory organ and urinary system Human ortholog(s) of this gene implicated in hypertension Orthologous to human F11R (F11 receptor)
Catalog No. 50-000-05639 Supplier ABclonal Technology Supplier No. A23909PM20UL
Predicted to enable PDZ domain binding activity integrin binding activity and protein homodimerization activity Involved in intestinal absorption regulation of cytokine production and regulation of membrane permeability Acts upstream of or within cell adhesion and epithelial cell differentiation Located in bicellular tight junction Is expressed in several structures including 4-8 cell stage embryo alimentary system respiratory system sensory organ and urinary system Human ortholog(s) of this gene implicated in hypertension Orthologous to human F11R (F11 receptor)