Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JAM-A/CD321 Human 50ug
SDP

Catalog No. 5000197100
Encompass_Preferred

Recombinant human JAM-A (CD321) extracellular fragment, expressed in HEK293 cells and purified with a C-terminal 6xHis tag. Supplied as a lyophilized powder from a filtered Tris-buffered solution, it is intended for biochemical assays, antibody validation, structural studies, and cell biology investigations of tight junction assembly and cell polarity.

  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • C-terminal 6xHis tag for straightforward detection and purification.
  • Sequence corresponds to residues S28-V238, representing the extracellular region.
  • Predicted molecular weight 27-31 kDa and purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation from 20 mM Tris-HCl, 150 mM NaCl, 100 mM glycine, pH 7.5.
  • Available in small unit sizes suitable for assay development and antibody validation.

Catalog No. 50-001-97100 Supplier Medchemexpress LLC Supplier No. HYP7094850UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human JAM-A (CD321) extracellular fragment, expressed in HEK293 cells and purified with a C-terminal 6xHis tag. Supplied as a lyophilized powder from a filtered Tris-buffered solution, it is intended for biochemical assays, antibody validation, structural studies, and cell biology investigations of tight junction assembly and cell polarity.

  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • C-terminal 6xHis tag for straightforward detection and purification.
  • Sequence corresponds to residues S28-V238, representing the extracellular region.
  • Predicted molecular weight 27-31 kDa and purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation from 20 mM Tris-HCl, 150 mM NaCl, 100 mM glycine, pH 7.5.
  • Available in small unit sizes suitable for assay development and antibody validation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.