Learn More
Aviva Systems Biology Corp JAM2 Antibody - middle region (ARP64440_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64440_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JAM2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human JAM2. Alternate names: C21orf43, CD322, JAM-B, JAMB, PRO245, VE-JAM, VEJAMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-263