Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Janelia Fluor 525, SE (JF525, SE) | 2975167-21-4 | ≥95.0% | 2 MG
SDP

Catalog No. 5000338994
Encompass_Preferred

Janelia Fluor 525, SE is a yellow fluorescent dye with an excitation wavelength of 525 nm and an emission wavelength of 549 nm. It contains an NHS ester, making it suitable for labeling primary amines and live cell imaging. This product is for research use only.

  • Yellow fluorescent dye
  • Excitation wavelength of 525 nm
  • Emission wavelength of 549 nm
  • Contains an NHS ester
  • Suitable for labeling primary amines
  • Suitable for live cell imaging

Catalog No. 50-003-38994 Supplier Medchemexpress LLC Supplier No. HY1310202MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Janelia Fluor 525, SE is a yellow fluorescent dye with an excitation wavelength of 525 nm and an emission wavelength of 549 nm. It contains an NHS ester, making it suitable for labeling primary amines and live cell imaging. This product is for research use only.

  • Yellow fluorescent dye
  • Excitation wavelength of 525 nm
  • Emission wavelength of 549 nm
  • Contains an NHS ester
  • Suitable for labeling primary amines
  • Suitable for live cell imaging

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.