Learn More
Medchemexpress LLC Janelia Fluor 646, SE (NHS ester) | 1811539-59-9 | 99.7% | 593.7 g/mol | C33H31N3O6Si | 5 MG
SDP
Catalog No. 5000262395
Encompass_Preferred
Janelia Fluor 646, SE is a far-red succinimidyl ester dye for covalent labeling of primary amines, designed for fluorescence microscopy and live-cell applications. The reagent provides bright, photostable far-red emission and is formulated for reliable conjugation and imaging performance.
  • Bright far-red fluorescence with excitation ~646 nm and emission ~664 nm.
  • High photostability for prolonged imaging sessions.
  • Reactive succinimidyl ester for efficient amine conjugation.
  • Suitable for confocal, super-resolution, and live-cell imaging.
  • High chemical purity for consistent labeling results.
Catalog No. 50-002-62395 Supplier Medchemexpress LLC Supplier No. HY1307355MG
Janelia Fluor 646, SE is a far-red succinimidyl ester dye for covalent labeling of primary amines, designed for fluorescence microscopy and live-cell applications. The reagent provides bright, photostable far-red emission and is formulated for reliable conjugation and imaging performance.
  • Bright far-red fluorescence with excitation ~646 nm and emission ~664 nm.
  • High photostability for prolonged imaging sessions.
  • Reactive succinimidyl ester for efficient amine conjugation.
  • Suitable for confocal, super-resolution, and live-cell imaging.
  • High chemical purity for consistent labeling results.