Learn More
Medchemexpress LLC Janelia Fluor 646, Tetrazine | 2042192-00-5 | 90.17% | 665.82 | 200 UG
SDP
Catalog No. 5000350790
Encompass_Preferred
Janelia Fluor 646, Tetrazine is a red fluorescent dye containing a tetrazine group. It is utilized in cellular imaging as a click chemistry reagent, capable of undergoing an inverse electron demand Diels-Alder reaction (iEDDA) with molecules that contain TCO groups.
  • Red fluorescent dye
  • Contains a tetrazine group
  • Suitable for cellular imaging
  • Click chemistry reagent
  • Undergoes inverse electron demand Diels-Alder reaction (iEDDA)
  • Photostable
  • Membrane-permeable
  • High extinction coefficient
Catalog No. 50-003-50790 Supplier Medchemexpress LLC Supplier No. HY138659200UG
Janelia Fluor 646, Tetrazine is a red fluorescent dye containing a tetrazine group. It is utilized in cellular imaging as a click chemistry reagent, capable of undergoing an inverse electron demand Diels-Alder reaction (iEDDA) with molecules that contain TCO groups.
  • Red fluorescent dye
  • Contains a tetrazine group
  • Suitable for cellular imaging
  • Click chemistry reagent
  • Undergoes inverse electron demand Diels-Alder reaction (iEDDA)
  • Photostable
  • Membrane-permeable
  • High extinction coefficient