Learn More
Medchemexpress LLC HY-15508 5mg Medchemexpress, JANEX-1 CAS:202475-60-3 Purity:>98%
SDP
Catalog No. 502030638
Encompass_Preferred
Medchemexpress, HY-15508 5mg JANEX-1 CAS:202475-60-3 JANEX-1 (WHI-P131) is a potent and specific JAK3 inhibitor (estimated Ki=2.3 μM). JANEX-1 (WHI-P131) shows potent JAK3-inhibitory activity (IC50 of 78 μM), does not inhibit JAK1 and JAK2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0638 Supplier Medchemexpress LLC Supplier No. HY155085MG
Medchemexpress, HY-15508 5mg JANEX-1 CAS:202475-60-3 JANEX-1 (WHI-P131) is a potent and specific JAK3 inhibitor (estimated Ki=2.3 μM). JANEX-1 (WHI-P131) shows potent JAK3-inhibitory activity (IC50 of 78 μM), does not inhibit JAK1 and JAK2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.