Learn More
Fluotics Janus comp P235 non-sterile
Catalog No. NC2196005
Encompass
PerkinElmer Janus compatible P235, non-sterile, for MDT head, working volume: 235 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 6001295
This item is not returnable. View return policy
Catalog No. NC2196005 Supplier Fluotics Supplier No. PKE235.NS
PerkinElmer Janus compatible P235, non-sterile, for MDT head, working volume: 235 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 6001295