Learn More
Fluotics Janus comp P30 non-sterile
Catalog No. NC2196007
Encompass_Preferred
PerkinElmer Janus compatible P30, non-sterile, for MDT head, working volume: 30 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 6001621
This item is not returnable. View return policy
Catalog No. NC2196007 Supplier Fluotics Supplier No. PKE30.NS
PerkinElmer Janus compatible P30, non-sterile, for MDT head, working volume: 30 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 6001621