Learn More
Description
- Clearly visualize the mitochondria of cells in histological tissue samples with this 1% aqueous janus green solution.
Specifications
Specifications
| Melting Point | ~0°C (32°F in water) |
| Boiling Point | 100°C |
| CAS Max % | 99%, 1% |
| CAS Min % | 99%, 1% |
| Concentration | 1% |
| Dye Content | 1% |
| Extinction Coefficient | 35000 M-1cm-1 (at 587 nm) |
| Identification | Janus Green B |
| Infrared Spectrum | Yes |
| Lambda Max | 587 nm |
| Show More |
Safety and Handling
| Hazard Category | May be harmful if swallowed. |
| ShelfLife | 3 years |
| DOTInformation | Not regulated. |
| EINECSNumber | 220-695-6 |
| Recommended Storage | Store in a cool, dry, well-ventilated area away from incompatible substances. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
