Learn More
Medchemexpress LLC Janus green B | 2869-83-2 | 98.75% | 511.06 | 1 G
SDP
Catalog No. 5000431343
Encompass_Preferred
Janus green B is a supravital stain with an oxygen-dependent staining reaction that is reversibly inhibited by cyanide. It is commonly used for staining peripheral nerves in live insects, lymphatic vessels of rabbits, and mitochondria. It is also suitable for observing new linear structures inside rabbit lymphatic vessels.
  • Used as a supravital stain
  • Suitable for observing new linear structures inside rabbit lymphatic vessels
  • Staining reaction is oxygen-dependent
Catalog No. 50-004-31343 Supplier Medchemexpress LLC Supplier No. HYD11221G
Janus green B is a supravital stain with an oxygen-dependent staining reaction that is reversibly inhibited by cyanide. It is commonly used for staining peripheral nerves in live insects, lymphatic vessels of rabbits, and mitochondria. It is also suitable for observing new linear structures inside rabbit lymphatic vessels.
  • Used as a supravital stain
  • Suitable for observing new linear structures inside rabbit lymphatic vessels
  • Staining reaction is oxygen-dependent