Learn More
Medchemexpress LLC Janus green B | 2869-83-2 | 98.8% | 511.06 | 500 MG
SDP
Catalog No. 5000428169
Encompass_Preferred
Janus green B is a supravital stain whose staining reaction is oxygen dependent and reversibly inhibited by cyanide. It has been used for staining peripheral nerves in live insects, lymphatic vessels of rabbits, and mitochondria.
  • Used in cell staining analysis
  • Applied in biology analysis
  • Utilized for histological analysis
  • Functions as an organelle dye
  • Employed in microbiological analysis
  • Suitable for histochemistry
  • Acts as a mitochondria dye
  • Used as a bacteriological dye
Catalog No. 50-004-28169 Supplier Medchemexpress LLC Supplier No. HYD1122500MG
Janus green B is a supravital stain whose staining reaction is oxygen dependent and reversibly inhibited by cyanide. It has been used for staining peripheral nerves in live insects, lymphatic vessels of rabbits, and mitochondria.
  • Used in cell staining analysis
  • Applied in biology analysis
  • Utilized for histological analysis
  • Functions as an organelle dye
  • Employed in microbiological analysis
  • Suitable for histochemistry
  • Acts as a mitochondria dye
  • Used as a bacteriological dye