Learn More
Medchemexpress LLC Janus green B | 2869-83-2 | MFCD00011758 | 98.8% | 511.06 | 100 MG
SDP
Catalog No. 5000428277
Encompass_Preferred
Janus green B is a supravital stain known for its oxygen-dependent staining reaction, which is reversibly inhibited by cyanide.
  • Stains peripheral nerves in live insects
  • Stains lymphatic vessels of rabbits
  • Stains mitochondria
Catalog No. 50-004-28277 Supplier Medchemexpress LLC Supplier No. HYD1122100MG
Janus green B is a supravital stain known for its oxygen-dependent staining reaction, which is reversibly inhibited by cyanide.
  • Stains peripheral nerves in live insects
  • Stains lymphatic vessels of rabbits
  • Stains mitochondria