Learn More
Medchemexpress LLC Janus kinase 2/JAK2 5ug | 5ug
SDP
Catalog No. 5000194707
Encompass_Preferred
Janus kinase 2/JAK2 Protein Human (His) is a recombinant human JAK2 expressed in E coli with a His tag at the N-terminus JAK2 is a non-receptor tyrosine kinase[1][2]
Catalog No. 50-001-94707 Supplier Medchemexpress LLC Supplier No. HYP79895UG
Janus kinase 2/JAK2 Protein Human (His) is a recombinant human JAK2 expressed in E coli with a His tag at the N-terminus JAK2 is a non-receptor tyrosine kinase[1][2]