Learn More
Bioss Japanese Encephalitis virus
SDP
Supplier:  Bioss BSM72275M
Encompass_Preferred
Japanese Encephalitis virus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65502