Learn More
Anton Paar Set of 2 tungsten carbide jars 25 ml
Catalog No. NC1818291
Encompass_Preferred
Grinding jar with 25 mL volume made of tungsten carbide. The perfect solution for materials like clinker, metal oxides, minerals, etc.Suitable for use with Anton Paar BM500 Ball Mill NC1818284
Catalog No. NC1818291 Supplier Anton Paar Supplier No. 177684
Add to Cart
Add to Cart
Request a Quote
Grinding jar with 25 mL volume made of tungsten carbide. The perfect solution for materials like clinker, metal oxides, minerals, etc.Suitable for use with Anton Paar BM500 Ball Mill NC1818284