Learn More
Anton Paar Set of 2 PTFE jars 50 ml
Catalog No. NC1818296
Encompass_Preferred
Grinding jar with 50 mL volume made of PTFE. The perfect solution for materials like drugs, biology, pharmaceuticals, plants, etc.Suitable for use with Anton Paar BM500 Ball Mill NC1818284
Catalog No. NC1818296 Supplier Anton Paar Supplier No. 177689
Grinding jar with 50 mL volume made of PTFE. The perfect solution for materials like drugs, biology, pharmaceuticals, plants, etc.Suitable for use with Anton Paar BM500 Ball Mill NC1818284