Learn More
Volu Sol Coplin Jar, Plastic
SDP
Supplier:  Volu Sol VCJ001
Encompass_Preferred
High-quality, plastic, coplin staining jars. Screw-on cover prevents evaporation, spills, and contamination when staining. Each jar holds five single slides or ten slides back to back. 1cs=12ea
Catalog No. NC9098929