Learn More
Newcomer Supply EasyDip Anodized Aluminum Jar Rack Holder
SDP
Catalog No. NC0795693
Encompass
The EasyDip Anodized Aluminum Jar Rack Holder helps prevent tipping and provides easy mobility of stationed jars if needed. It is made of anodized aluminum to resist rust, corrosion and abarasion and holds up to six EasyDip Slide Staining Jars. Temperature range: -80°C to +30°C. Dimensions: 16 3/4" x 4" x 1 1/2" (H).
Catalog No. NC0795693 Supplier Newcomer Supply Supplier No. 5300JRK
The EasyDip Anodized Aluminum Jar Rack Holder helps prevent tipping and provides easy mobility of stationed jars if needed. It is made of anodized aluminum to resist rust, corrosion and abarasion and holds up to six EasyDip Slide Staining Jars. Temperature range: -80°C to +30°C. Dimensions: 16 3/4" x 4" x 1 1/2" (H).