Learn More
BPS Bioscience Inc JARID1A (KDM5A) Homogeneous Assay Kit, 384 reactions
SDP
Catalog No. NC4036028
Encompass
The JARID1A (KDM5A) Homogenous Assay Kit is designed to measure activity of the JARID1A (Jumonji AT-rich interactive domain 1A, also known as KDM5A) for screening and profiling purposes. The kit comes in a convenient format 96-well AlphaLISA® format, with enough purified recombinant JARID1A (amino acids 1-1090), primary antibody, substrate and buffers for 100 enzyme reactions.
Catalog No. NC4036028 Supplier BPS Bioscience Inc Supplier No. 505102
The JARID1A (KDM5A) Homogenous Assay Kit is designed to measure activity of the JARID1A (Jumonji AT-rich interactive domain 1A, also known as KDM5A) for screening and profiling purposes. The kit comes in a convenient format 96-well AlphaLISA® format, with enough purified recombinant JARID1A (amino acids 1-1090), primary antibody, substrate and buffers for 100 enzyme reactions.