Learn More
Cayman Chemical JARID1BPLU1C-Term Polyclo 1ea
SDP
Catalog No. 502969726
Encompass_Preferred
Immunogen: Synthetic peptide from the C-terminal region of human JARID1B/PLU1 • Host: rabbit • Cross Reactivity: (+) human JARID1B/PLU1 • Application(s): FC and ICC
Catalog No. 50-296-9726 Supplier Cayman Chemical Supplier No. 147011 EA
Immunogen: Synthetic peptide from the C-terminal region of human JARID1B/PLU1 • Host: rabbit • Cross Reactivity: (+) human JARID1B/PLU1 • Application(s): FC and ICC