Learn More
Apexbio Technology LLC (±)-Jasmonic Acid methyl ester 39924-52-2 25g
SDP
Supplier:  Apexbio Technology LLC C440325000
Encompass_Preferred
( )-Jasmonic Acid methyl ester (CAS 39924-52-2) is a plant-derived compound belonging to the jasmonate family of stress hormones It modulates cellular processes by inducing the accumulation of proteinase inhibitors in plant tissues and can trigger apoptotic pathways in mammalian cancer cell lines characterized by altered nuclear morphology and elevated caspase-3 activity In plant studies exposure to ( )-Jasmonic Acid methyl ester led to enhanced defensive responses In vitro experiments demonstrated its capacity to suppress proliferation and induce apoptosis in various tumor cell lines without affecting normal lymphocytes In vivo treatment prolonged survival in mouse lymphoma models This compound is used to investigate stress response signaling and apoptosis mechanisms in both plant and mammalian systems
Catalog No. 50-000-88285